Anti-TLK1 Rabbit Polyclonal Antibody
ROCK600-401-469
- Antibody type:Primary
- Antigen name:Tousled-like Kinase 1
- Antigen symbol:TLK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Orangutan,Chimpanzee,Human,Dog
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:9874
- Antigen synonyms:PKU-beta
- Amino acid number:737 to 751
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,60 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 737-751 of human Tlk1 (Tousled-like kinase) (isoform 1) Protein.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
This affinity purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 86 kDa, 89 kDa or 64 kDa in size corresponding to isoforms of TLk-1.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:200 - 1:2000;
Type: Primary
Antigen: TLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Orangutan, Dog