Anti-NFKB cRel Rabbit Polyclonal Antibody
ROCK100-4166
- Antibody type:Primary
- Antigen name:Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
- Antigen symbol:NFKB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:near the C-terminus
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:4790
- Antigen synonyms:NFkappaB|EBP-1|p50|NFKB-p50|NFKB-p105|NF-kB1|NF-kappaB|p105|KBF1|NF-kappa-B
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:80 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:NFkB cRel peptide corresponding to a region near the C-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
This product was assayed by immunoblot and found to be reactive against NFkB cRel at a dilution of 1:1000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).
Recommended Dilutions: ELISA: 1:5000 - 1:25000; GelShift: 1:500; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: NFKB cRel
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype:
Reactivity: Human