Anti-Elastase, Neutrophil Mouse monoclonal antibody unconjugated
Catalog # QEDB13203
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Elastase, Neutrophil
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:none
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Elastase, Neutrophil
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human