Anti-SLIMMER / FHL1 isoform B Goat Polyclonal Antibody
EVBIEB06508
- Antibody type:Primary
- Antigen name:FHL1 isoform B
- Antigen symbol:SLIMMER
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: SLIMMER / FHL1 isoform B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse