Order Entry
Austria
ContactUsLinkComponent
Anti-PU.1/Spi1 Rabbit Polyclonal Antibody
Anti-PU.1/Spi1 Rabbit Polyclonal Antibody
  NOVUNBP2-27163
undefined
Anti-PU.1/Spi1 Rabbit Polyclonal Antibody
  NOVUNBP2-27163

 

  • Antibody type:
    Primary
  • Antigen name:
    Spi-1 proto-oncogene
  • Antigen symbol:
    SPI1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    6688
  • Antigen synonyms:
    SPI-A|SPI-1|PU.1|SFPI1|OF
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: PU.1/Spi1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human