Anti-HOMER1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: HOMER1 (homer homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-44999
- NOVUNBP1-44999SS
- Antigen symbol:
- HOMER1
- HOMER1
- Antigen name:
- Homer protein homolog 1
- Homer protein homolog 1
- Antigen synonyms:
- HOMER|Ves-1|SYN47|HOMER1B|HOMER1A|HOMER1C
- HOMER|Ves-1|SYN47|HOMER1B|HOMER1A|HOMER1C
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 9456
- 9456
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together