Order Entry
Austria
ContactUsLinkComponent
 
Anti-SCP1 Rabbit Polyclonal Antibody
 
undefined
Anti-SCP1 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB300-229
  • NOVUNB300-229SS
  • Antigen symbol:
  • SCP1
  • SCP1
  • Antigen name:
  • Serine carboxypeptidase 1
  • Serine carboxypeptidase 1
  • Antigen synonyms:
  • RISC|SCP1|Retinoid-inducible serine carboxypeptidase|SCPEP1|UNQ265/PRO302|Serine carboxypeptidase 1|MSTP034|HSCP1
  • RISC|SCP1|Retinoid-inducible serine carboxypeptidase|SCPEP1|UNQ265/PRO302|Serine carboxypeptidase 1|MSTP034|HSCP1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6847
  • 6847
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together