Anti-Peroxiredoxin 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1837
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Peroxiredoxin 4
- Antigen symbol:PRDX4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:PRX-4|Peroxiredoxin-4|PRX 4|Thioredoxin Peroxidase AO372|Thioredoxin-Dependent Peroxide Reductase A0372|PRX4|prx-IV|Antioxidant enzyme AOE372|Peroxiredoxin IV|PRX IV|Thioredoxin Peroxidase (antioxidant enzyme)|AOE37-2
- Amino acid number:253 - 271
- Sequence:SETIIPDPAGKLKYFDKLN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Peroxiredoxin 4 (253-271aa SETIIPDPAGKLKYFDKLN), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Peroxiredoxin-4 (PRDX4) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Peroxiredoxin 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat