Order Entry
Austria
ContactUsLinkComponent
 
Anti-CPT1A Mouse Monoclonal Antibody [clone: 8F6AE9]
  ABCAAB128568-100
 :  Abcam
Anti-CPT1A Mouse Monoclonal Antibody [clone: 8F6AE9]
  ABCAAB128568-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    carnitine palmitoyltransferase 1A (liver)
  • Antigen symbol:
    CPT1A
  • Clonality:
    Monoclonal
  • Clone:
    8F6AE9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CPT1A (carnitine palmitoyltransferase 1A (liver))
Clonality: monoclonal
Clone: 8F6AE9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: