Order Entry
Austria
ContactUsLinkComponent
Anti-ZIP Kinase Rabbit Monoclonal Antibody [clone: EPR1636Y]
Anti-ZIP Kinase Rabbit Monoclonal Antibody [clone: EPR1636Y]
  ABCAAB51602-100
 :  Abcam
Anti-ZIP Kinase Rabbit Monoclonal Antibody [clone: EPR1636Y]
  ABCAAB51602-100
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen symbol:
    ZIP Kinase
  • Clonality:
    Monoclonal
  • Clone:
    EPR1636Y
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ZIP Kinase
Clonality: monoclonal
Clone: EPR1636Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: