Anti-GLG1 Rabbit Polyclonal Antibody
Catalog # BIRBORB96724-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Golgi glycoprotein 1
- Antigen symbol:GLG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic 18 amino acid peptide from internal region of human GLG1
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Immunochemistry: 1.25-2.5 ?g/ml
Type: Primary
Antigen: GLG1 (golgi glycoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: