Anti-CREB3L1 Goat Polyclonal Antibody
Catalog # BIRBORB89595-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cAMP responsive element binding Protein 3-like 1
- Antigen symbol:CREB3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
ELISA: 1:32000; Immunochemistry: 3.75 ?g/ml; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: CREB3L1 (cAMP responsive element binding protein 3-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat