Anti-NCK beta Rabbit Polyclonal Antibody
BIRBORB87260-50
: Biorbyt
- Antibody type:Primary
- Antigen name:NCK beta
- Antigen symbol:NCKBETA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Storage buffer:PBS, pH 7,4, 0,2% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:0,2 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: NCK beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human