Anti-GST tag Mouse Monoclonal Antibody
Supplier: Biorbyt
Certificates
About this item
- Western Blot: 1:3000-1:10000
Type: Primary
Antigen: GST tag (Glutathione-S-Transferase tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Specifications
- Catalog No:
- BIRBORB66651-20
- BIRBORB66651-100
- Antigen symbol:
- GST
- GST
- Antigen name:
- Glutathione-S-Transferase
- Glutathione-S-Transferase
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- Immunogen:
- Full length GST protein
- Full length GST protein
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- 20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Storage buffer:
- Mouse IgG in phosphate buffered saline, pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Mouse IgG in phosphate buffered saline, pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together