Anti-calcium binding protein P22 Rabbit Polyclonal Antibody
Catalog # BIRBORB41706-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:calcium binding Protein P22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Amino acid number:47 to 111
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide contain a sequence corresponding to a region within amino acids 47 and 111 of human CHP.
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000
Type: Primary
Antigen: calcium binding protein P22
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human