Anti-PIN-Cocktail 1 P504S / p63 None Monoclonal Antibody
: Biorbyt
Type: Primary
Antigen: PIN-Cocktail 1 P504S / p63
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG+IgG2a
Reactivity: Human
- Catalog No:
- BIRBORB27516-0.1
- BIRBORB27516-1
- BIRBORB27516-0.5
- Antigen symbol:
- P63
- P63
- P63
- Antigen name:
- PIN-Cocktail 1 P504S
- PIN-Cocktail 1 P504S
- PIN-Cocktail 1 P504S
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Isotype:
- IgG, IgG2a
- IgG, IgG2a
- IgG, IgG2a
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Immunogen:
- Human recombinant AMACR (P504S) protein, whole sequence
- Human recombinant AMACR (P504S) protein, whole sequence
- Format:
- concentrated
- concentrated
- Form:
- liquid
- liquid
- Concentration:
- 0.0694444444
- 0.0694444444
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
Frequently Bought Together