Anti-RAX2 Mouse Polyclonal Antibody
Catalog # ABNOH00084839-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retina and anterior neural fold homeobox 2
- Antigen symbol:RAX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:84839
- Antigen synonyms:ARMD6, CORD11, QRX, RAXL1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: RAX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human