Anti-GPR84 Mouse Monoclonal Antibody [clone: 5B7]
Catalog # ABNOH00053831-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 84
- Antigen symbol:GPR84
- Clonality:Monoclonal
- Clone:5B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:53831
- Antigen synonyms:GPCR4|EX33
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GPR84
Clonality: Monoclonal
Clone: 5B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human