Anti-ZAK Mouse Monoclonal Antibody [clone: 3G5]
ABNOH00051776-M03A
: Abnova
- Antibody type:Primary
- Antigen name:sterile alpha motif and leucine zipper containing kinase AZK
- Antigen symbol:ZAK
- Clonality:Monoclonal
- Clone:3G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51776
- Antigen synonyms:MLTK|pk|MRK|MLK7|mlklak|AZK|MLT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: ZAK
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human