Anti-CNOT7 Mouse Monoclonal Antibody [clone: 2F6]
Catalog # ABNOH00029883-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CCR4-NOT transcription complex, subunit 7
- Antigen symbol:CNOT7
- Clonality:Monoclonal
- Clone:2F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:29883
- Antigen synonyms:hCAF-1|Caf1a|CAF1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CNOT7
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat