Anti-GCA Mouse Monoclonal Antibody [clone: 3D12]
ABNOH00025801-M02
: Abnova
- Antibody type:Primary
- Antigen name:Grancalcin, EF-hand Calcium Binding Protein
- Antigen symbol:GCA
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:25801
- Antigen synonyms:GCL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GCA
Clonality: Monoclonal
Clone: 3D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human