Anti-TRIM31 Mouse Polyclonal Antibody
ABNOH00011074-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Tripartite Motif Containing 31
- Antigen symbol:TRIM31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11074
- Antigen synonyms:RNF|HCGI|C6orf13|Tripartite mot containing protein 31|H.sapiens mRNA for RING protein|Ring finger protein|HCG1|TRI31_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: TRIM31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human