Anti-TTR Mouse Monoclonal Antibody [clone: 4D8]
ABNOH00007276-M01
: Abnova
- Antibody type:Primary
- Antigen name:Transthyretin
- Antigen symbol:TTR
- Clonality:Monoclonal
- Clone:4D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7276
- Antigen synonyms:HEL111|CTS1|TBPA|HsT2651|Transthyretin|CTS|PALB|ATTR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TTR
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human