Anti-LEFTY2 Mouse Monoclonal Antibody [clone: 1H6]
Catalog # ABNOH00007044-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Left-right determination Factor 2
- Antigen symbol:LEFTY2
- Clonality:Monoclonal
- Clone:1H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7044
- Antigen synonyms:TGFB4|LEFTA|LEFTYA|EBAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LEFTY2
Clonality: Monoclonal
Clone: 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human