Anti-SLC9A1 Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00006548-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 9 Member 1
- Antigen symbol:Solute carrier family 9, member 1
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6548
- Antigen synonyms:NHE-1|PPP1R143|Sodiumûhydrogen antiporter 1|APNH|solute carrier family 9 member A1|NHE1|LIKNS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLC9A1
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human