Anti-CCL23 Rabbit Polyclonal Antibody
ABNOH00006368-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 23
- Antigen symbol:CCL23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6368
- Antigen synonyms:hmrp-2a|MIP3|CK-BETA-8|SCYA23|MPIF-1|MIP-3|Ckb-8-1|Ckb-8|CKb8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CCL23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human