Anti-RPLP0 Mouse Monoclonal Antibody [clone: 1B4]
Catalog # ABNOH00006175-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein Large P0
- Antigen symbol:RPLP0
- Clonality:Monoclonal
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6175
- Antigen synonyms:P0|RPP0|60S acidic ribosomal protein P0|PRLP0|L10E|RPLP0|60S ribosomal protein L10E|LP0
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RPLP0
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human