Anti-RPL7 Mouse Monoclonal Antibody [clone: 2E10]
ABNOH00006129-M06
: Abnova
- Antibody type:Primary
- Antigen name:Ribosomal Protein L7
- Antigen symbol:RPL7
- Clonality:Monoclonal
- Clone:2E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6129
- Antigen synonyms:humL7-1|L7|60S ribosomal protein L7|RPL7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RPL7
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human