Anti-RCN2 Mouse Polyclonal Antibody
ABNOH00005955-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Reticulocalbin 2
- Antigen symbol:RCN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5955
- Antigen synonyms:E6BP|TCBP49|ERC-55|ERC55
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: RCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human