Anti-MED1 Mouse Monoclonal Antibody [clone: 1G3]
Catalog # ABNOH00005469-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mediator Complex Subunit 1
- Antigen symbol:MED1
- Clonality:Monoclonal
- Clone:1G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5469
- Antigen synonyms:TRIP-2|Activator-recruited cofactor 205 kDa component|TRIP2|PPAR-binding protein|PPARBP|Peroxisome proliferator-activated receptor-binding protein|ARC205|Mediator of RNA polymerase II transcription subunit 1|PBP|Mediator complex subunit 1|Trap220|Thyroid hormone receptor-associated protein complex 220 kDa component|TR-interacting protein 2|DRIP230|CRSP200|MED1|PPARGBP|Thyroid receptor-interacting protein 2|DRIP205|CRSP1|Vitamin D receptor-interacting protein complex component DRIP205|p53 regulatory protein RB18A|TRAP220|RB18A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MED1
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human