Anti-PON3 Mouse Polyclonal Antibody
ABNOH00005446-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Paraoxonase 3
- Antigen symbol:PON3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5446
- Antigen synonyms:PON3_HUMAN|MGC95026.|AI786302|Pon3|2810004E20|Serum paraoxonase/lactonase 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PON3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human