Anti-MPP1 Mouse Monoclonal Antibody [clone: 1E11-1G11]
ABNOH00004354-M01
: Abnova
- Antibody type:Primary
- Antigen name:Membrane Palmitoylated Protein 1
- Antigen symbol:MPP1
- Clonality:Monoclonal
- Clone:1E11-1G11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4354
- Antigen synonyms:DXS552E|AAG12|MRG1|55 kDa erythrocyte membrane protein|PEMP|EMP55
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MPP1
Clonality: Monoclonal
Clone: 1E11-1G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human