Anti-CK35 Rabbit Polyclonal Antibody
Catalog # ABNOH00003886-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 35
- Antigen symbol:CK35
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3886
- Antigen synonyms:Ha-5|hHa5|KRTHA5|HA5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CK35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human