Order Entry
Austria
ContactUsLinkComponent
Anti-HMX2 Mouse Monoclonal Antibody [clone: 2D2]
Anti-HMX2 Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ABNOH00003167-M09
Supplier:  Abnova
undefined
Anti-HMX2 Mouse Monoclonal Antibody [clone: 2D2]
Catalog # ABNOH00003167-M09
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    H6 Homeobox 2
  • Antigen symbol:
    HMX2
  • Clonality:
    Monoclonal
  • Clone:
    2D2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    3167
  • Antigen synonyms:
    H6L|Nkx5-2
  • Storage buffer:
    In 1x PBS, pH 7,4
  • Sequence:
    PAGSPSPGSERPRDGGAERQAGAAKKKTRTVFSRSQVYQLESTFDMKRYLSSSERACLASSLQLTETQVKTWFQNRRNKWKRQLSAELEAANMAHASAQT
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Shipping temperature:
    Ice
  • Immunogen:
    HMX2 (NP_005510, 125 a.a. ~ 224 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a full length recombinant HMX2.

Recommended Dilutions: Western Blot (1 - 5 µg/ml)

Type: Primary
Antigen: HMX2
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human