Anti-HLA-DQB2 Mouse Polyclonal Antibody
ABNOH00003120-B01P
: Abnova
- Antibody type:Primary
- Antigen name:major histocompatibility complex, class II, DQ beta 2
- Antigen symbol:HLA-DQB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3120
- Antigen synonyms:HLA-DQB1|HLA-DXB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: HLA-DQB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human