Anti-GOLGA2 Mouse Monoclonal Antibody [clone: 2C6]
Catalog # ABNOH00002801-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Golgin A2
- Antigen symbol:GOLGA2
- Clonality:Monoclonal
- Clone:2C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2801
- Antigen synonyms:GM130
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GOLGA2
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human