Order Entry
Austria
ContactUsLinkComponent
 
Anti-FOSB Rabbit Polyclonal Antibody
  ABNOH00002354-D01
 :  Abnova
undefined
Anti-FOSB Rabbit Polyclonal Antibody
  ABNOH00002354-D01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    FBJ murine osteosarcoma viral oncogene homolog B
  • Antigen symbol:
    FOSB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    2354
  • Antigen synonyms:
    AP-1|GOSB|G0S3|GOS3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: FOSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human