Order Entry
Austria
ContactUsLinkComponent
Anti-CTSC Mouse Polyclonal Antibody
Anti-CTSC Mouse Polyclonal Antibody
Catalog # ABNOH00001075-B01P
Supplier:  Abnova
undefined
Anti-CTSC Mouse Polyclonal Antibody
Catalog # ABNOH00001075-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin C
  • Antigen symbol:
    CTSC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1075
  • Antigen synonyms:
    JP|DPP1|DPP-I|JPD|HMS|PLS|CPPI|DPPI|PDON1|PALS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: CTSC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human