Anti-ALDOB Mouse Monoclonal Antibody [clone: 3B3]
ABNOH00000229-M01
: Abnova
- Antibody type:Primary
- Antigen name:Aldolase B, fructose-bisphosphate
- Antigen symbol:ALDOB
- Clonality:Monoclonal
- Clone:3B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:229
- Antigen synonyms:ALDO2|ALDB
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ALDOB
Clonality: Monoclonal
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human