Anti-CPT1C Goat Antibody
ORIGTA311581
: OriGene
- Antibody type:Primary
- Antigen name:carnitine palmitoyltransferase 1C
- Antigen symbol:CPT1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.Aliquot and store at -20C. Minimize freezing and thawing.
- Sequence:C-KSSTKTDSHRLGQH
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Pk:100 µG
Recommended Dilutions: Western Blot: 1-3?g/ml
Type: Primary
Antigen: CPT1C (carnitine palmitoyltransferase 1C)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human