Order Entry
Austria
ContactUsLinkComponent
Anti-CHKA Rabbit Polyclonal Antibody
Anti-CHKA Rabbit Polyclonal Antibody
Catalog # ABGEAP8179B
Supplier:  Abgent
undefined
Anti-CHKA Rabbit Polyclonal Antibody
Catalog # ABGEAP8179B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Choline kinase alpha
  • Antigen symbol:
    CHKA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Gene ID:
    12534
  • Antigen synonyms:
    CK|EK|CKI|CHK
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    CHPYFDDLDNQIKKM
  • Storage temperature:
    Cool
  • Concentration:
    0,393 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    This Mouse Cdk1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 269-297 amino acids from the C-terminal region of mouse Cdk1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CHKA (choline kinase alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human