Anti-GMFG Rabbit Polyclonal Antibody
ABGEAP7871A
: Abgent
- Antibody type:Primary
- Antigen name:Glia Maturation Factor Gamma
- Antigen symbol:GMFG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:GMF-gamma|GMFG|GMFG_HUMAN|Glia maturation factor gamma|MGC126867
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GMFG (glia maturation factor, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human