Anti-CTGF Rabbit Polyclonal Antibody
ABGEAP7778B
: Abgent
- Antibody type:Primary
- Antigen name:Connective Tissue Growth Factor
- Antigen symbol:CTGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:Insulin-like Growth Factor-Binding Protein 8,|IGFBP8|Hcs24|CCN2|Hypertrophic chondrocyte-specific gene product 24|Connective Tissue Growth Factor
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human