Order Entry
Austria
ContactUsLinkComponent
 
Anti-RASGRF1 Rabbit Polyclonal Antibody
  ABGEAP7582B
 :  Abgent
undefined
Anti-RASGRF1 Rabbit Polyclonal Antibody
  ABGEAP7582B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Ras protein-specific guanine nucleotide-releasing factor 1
  • Antigen symbol:
    RASGRF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Antigen synonyms:
    CDC25|H-GRF55|KIAA0771|GRF55|CDC25L|ASPP1|Protein phosphatase 1 regulatory subunit 13B|PPP1R13B|ras-GRF1|Apoptosis-stimulating of p53 protein 1|GRF1|GNRP|PP13187
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RASGRF1 (Ras protein-specific guanine nucleotide-releasing factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human