Anti-FARSA Rabbit Polyclonal Antibody
Catalog # ABGEAP7566A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Phenylalanyl-tRNA Synthetase, Alpha subunit
- Antigen symbol:FARSA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:CML33|FARSA|Phenylalanyl-tRNA synthetase alpha chain|FARS|PheRS|Phenylalanine--tRNA ligase alpha chain|FARSLA|FARSL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: FARSA (phenylalanyl-tRNA synthetase, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human