Order Entry
Austria
ContactUsLinkComponent
Anti-DYRK2 Rabbit Polyclonal Antibody
Anti-DYRK2 Rabbit Polyclonal Antibody
  ABGEAP7534A
 :  Abgent
undefined
Anti-DYRK2 Rabbit Polyclonal Antibody
  ABGEAP7534A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2
  • Antigen symbol:
    DYRK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-Terminal (1–100 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    D6S230E|FPPR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: DYRK2 (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human