Order Entry
Austria
ContactUsLinkComponent
Anti-CEACAM Rabbit Polyclonal Antibody
Anti-CEACAM Rabbit Polyclonal Antibody
Catalog # ABGEAP7364A
Supplier:  Abgent
undefined
Anti-CEACAM Rabbit Polyclonal Antibody
Catalog # ABGEAP7364A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein)
  • Antigen symbol:
    CEACAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    Cc1|Bgp|Bgp1|Cea7|Mhv-1|Hv-2|CD66a|mmCGM1|Hv2|mmCGM2|Cea-1|mCEA1|C-CAM|MHVR|MHVR1|Cea-7|bb-1|Cea1|mmCGM1a
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CEACAM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human