Anti-GAB2 Rabbit Polyclonal Antibody
ABGEAP6908A
: Abgent
- Antibody type:Primary
- Antigen name:GRB2-associated binding protein 2
- Antigen symbol:GAB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Antigen synonyms:growth factor receptor bound protein 2-associated protein 2|Grb2-associated binder 2|GRB2-associated-binding protein 2|pp100
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: GAB2 (GRB2-associated binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human