Order Entry
Austria
ContactUsLinkComponent
Anti-SYN3 Rabbit Polyclonal Antibody
Anti-SYN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP5755C
Supplier:  Abgent
undefined
Anti-SYN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP5755C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synapsin 3
  • Antigen symbol:
    SYN3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    middle
  • Antigen synonyms:
    Synapsin III|Synapsin 3|Syn3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SYN3 (synapsin III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human