Order Entry
Austria
ContactUsLinkComponent
 
Anti-TNFAIP1 Rabbit Polyclonal Antibody
  ABGEAP5410B
 :  Abgent
undefined
Anti-TNFAIP1 Rabbit Polyclonal Antibody
  ABGEAP5410B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Alpha-induced Protein 1
  • Antigen symbol:
    TNFAIP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Antigen synonyms:
    EDP1|hBACURD2|B12|BTBD34|B61
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TNFAIP1 (tumor necrosis factor, alpha-induced protein 1 (endothelial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse